Learn More
MSC Vermont Gage Class ZZ Minus Plug & Pin Gage, 0.2130 in, 2 in
Catalog No. 5000534418
Encompass
This plug and pin gage is designed for accurate measurement and inspection in various applications. It offers precise dimensions and comes with a certificate of accuracy for reliable results.
  • Measures 0.2130 in diameter
  • Features an overall length of 2 in
  • Maintains a tight tolerance of 0.000200 in
  • Constructed from durable steel
  • Finished with black oxide coating for protection
  • Ideal for setting micrometers, Go/No-Go inspection, and calibrating verniers and other measuring devices
  • Includes a certificate of accuracy with NIST traceability
Catalog No. 50-005-34418 Supplier MSC Supplier No. 71832315
This plug and pin gage is designed for accurate measurement and inspection in various applications. It offers precise dimensions and comes with a certificate of accuracy for reliable results.
  • Measures 0.2130 in diameter
  • Features an overall length of 2 in
  • Maintains a tight tolerance of 0.000200 in
  • Constructed from durable steel
  • Finished with black oxide coating for protection
  • Ideal for setting micrometers, Go/No-Go inspection, and calibrating verniers and other measuring devices
  • Includes a certificate of accuracy with NIST traceability