Learn More
MSC Vermont Gage Class ZZ Minus Plug & Pin Gage, 0.2155 in Diameter, 2 in Overall Length, Steel, 10 Micro Finish
Catalog No. 5000511256
Encompass
This Class ZZ Minus Plug & Pin Gage is designed for precise measurement applications such as setting micrometers, performing NoGo inspections, and calibrating verniers and other measuring devices. It features a 0.2155 inch diameter and a 2 inch overall length, constructed from steel with a 10 micro finish. Each gage includes a Certificate of Accuracy with NIST Traceability.
  • Designed for setting micrometers, NoGo inspection, and calibrating measuring devices.
  • Features a 0.2155 inch diameter.
  • Has a 2 inch overall length.
  • Constructed from steel for durability.
  • Finished with a 10 micro coating for precision.
  • Includes a Certificate of Accuracy with NIST Traceability.
  • Offers a tolerance of 0.000200 inch for accurate measurements.
Catalog No. 50-005-11256 Supplier MSC Supplier No. 89021554
This Class ZZ Minus Plug & Pin Gage is designed for precise measurement applications such as setting micrometers, performing NoGo inspections, and calibrating verniers and other measuring devices. It features a 0.2155 inch diameter and a 2 inch overall length, constructed from steel with a 10 micro finish. Each gage includes a Certificate of Accuracy with NIST Traceability.
  • Designed for setting micrometers, NoGo inspection, and calibrating measuring devices.
  • Features a 0.2155 inch diameter.
  • Has a 2 inch overall length.
  • Constructed from steel for durability.
  • Finished with a 10 micro coating for precision.
  • Includes a Certificate of Accuracy with NIST Traceability.
  • Offers a tolerance of 0.000200 inch for accurate measurements.