Learn More
MSC Vermont Gage Class ZZ Minus Plug & Pin Gage, 0.2300 in Diameter, 2 in OAL, Steel
Catalog No. 5000515778
Encompass
This Class ZZ Minus Plug & Pin Gage features a 0.2300 in diameter and a 2 in overall length. It is constructed from steel with a 10 micro finish, suitable for precise measurement applications such as setting micrometers, verniers, and for NoGo inspection.
  • Certificate of accuracy with NIST traceability
  • Suitable for setting micrometers, verniers, and other measuring devices
  • Used for NoGo inspection
Catalog No. 50-005-15778 Supplier MSC Supplier No. 89023006
This Class ZZ Minus Plug & Pin Gage features a 0.2300 in diameter and a 2 in overall length. It is constructed from steel with a 10 micro finish, suitable for precise measurement applications such as setting micrometers, verniers, and for NoGo inspection.
  • Certificate of accuracy with NIST traceability
  • Suitable for setting micrometers, verniers, and other measuring devices
  • Used for NoGo inspection