Learn More
MSC Vermont Gage Class ZZ Minus Plug & Pin Gage, 0.2515 in, 2 in OAL
Catalog No. 5000530460
Encompass
This plug and pin gage is designed for precise measurement applications, including setting micrometers, verniers, and other measuring devices, as well as for NoGo inspection. Constructed from steel, it offers a tolerance of 0.000200 "
  • Set micrometers, verniers, and other measuring devices
  • Suitable for NoGo inspection
  • Includes a certificate of accuracy with NIST traceability
  • Features a 10 micro coating/finish
  • Constructed from durable steel
  • Offers a precise tolerance of 0.000200 inch
Catalog No. 50-005-30460 Supplier MSC Supplier No. 89025159
This plug and pin gage is designed for precise measurement applications, including setting micrometers, verniers, and other measuring devices, as well as for NoGo inspection. Constructed from steel, it offers a tolerance of 0.000200 "
  • Set micrometers, verniers, and other measuring devices
  • Suitable for NoGo inspection
  • Includes a certificate of accuracy with NIST traceability
  • Features a 10 micro coating/finish
  • Constructed from durable steel
  • Offers a precise tolerance of 0.000200 inch