Learn More
MSC Vermont Gage Class ZZ Minus Plug & Pin Gage, 0.2730 Inch Diameter, 2 Inch Length, Black Oxide
Catalog No. 5000528073
Encompass
This Vermont Gage Class ZZ Minus Plug & Pin Gage is precisely engineered for accurate measurement. Crafted from durable steel with a black oxide finish, it features a 0.2730 inch diameter and a 2 inch overall length. This gage is ideal for setting micrometers, for NoGo inspection, and for verifying verniers and other measuring devices.
  • Precision measurement with a diameter of 0.2730 "
  • Durable steel construction with a black oxide coating for enhanced longevity.
  • Includes a certificate of accuracy with NIST traceability.
  • Suitable for various inspection and calibration tasks.
  • Designed for a tolerance of 0.000200 "
  • Overall length of 2 inches.
Catalog No. 50-005-28073 Supplier MSC Supplier No. 60382645
This Vermont Gage Class ZZ Minus Plug & Pin Gage is precisely engineered for accurate measurement. Crafted from durable steel with a black oxide finish, it features a 0.2730 inch diameter and a 2 inch overall length. This gage is ideal for setting micrometers, for NoGo inspection, and for verifying verniers and other measuring devices.
  • Precision measurement with a diameter of 0.2730 "
  • Durable steel construction with a black oxide coating for enhanced longevity.
  • Includes a certificate of accuracy with NIST traceability.
  • Suitable for various inspection and calibration tasks.
  • Designed for a tolerance of 0.000200 "
  • Overall length of 2 inches.