Learn More
MSC Vermont Gage Class ZZ Minus Plug & Pin Gage, 0.3220 in Diameter, 2 in Overall Length, Steel
Catalog No. 5000514825
Encompass
The Class ZZ Minus Plug & Pin Gage is a precision tool crafted from steel, designed for accurate measurement and inspection tasks. With a diameter of 0.3220 inches and an overall length of 2 inches, it is suitable for setting micrometers, Go/NoGo inspection, and calibrating verniers and other measuring devices.
  • Designed for precise measurement and inspection tasks
  • Crafted from durable steel
  • Features a 10 micro coating/finish for enhanced performance
  • Includes a certificate of accuracy with NIST traceability, ensuring reliability
  • Ideal for setting micrometers
  • Suitable for NoGo inspection
  • Useful for calibrating verniers and other measuring devices
Catalog No. 50-005-14825 Supplier MSC Supplier No. 89032205
The Class ZZ Minus Plug & Pin Gage is a precision tool crafted from steel, designed for accurate measurement and inspection tasks. With a diameter of 0.3220 inches and an overall length of 2 inches, it is suitable for setting micrometers, Go/NoGo inspection, and calibrating verniers and other measuring devices.
  • Designed for precise measurement and inspection tasks
  • Crafted from durable steel
  • Features a 10 micro coating/finish for enhanced performance
  • Includes a certificate of accuracy with NIST traceability, ensuring reliability
  • Ideal for setting micrometers
  • Suitable for NoGo inspection
  • Useful for calibrating verniers and other measuring devices