Learn More
MSC Vermont Gage Class ZZ Minus Plug & Pin Gage, 0.3270 in Diameter, 2 in Overall Length, Steel
Catalog No. 5000531462
Encompass
The Vermont Gage Class ZZ Minus Plug & Pin Gage is a precise tool designed for critical measurement applications. Made from durable steel, it features a 0.3270 inch diameter and a 2 inch overall length. This gage is ideal for setting micrometers, NoGo inspection, and verifying other measuring devices.
  • Coating/finish: 10 Micro
  • Includes certificate of accuracy with NIST traceability
  • Suitable for setting micrometers, NoGo inspection, verniers, and other measuring devices
  • Tolerance of 0.000200 inch
Catalog No. 50-005-31462 Supplier MSC Supplier No. 89032700
The Vermont Gage Class ZZ Minus Plug & Pin Gage is a precise tool designed for critical measurement applications. Made from durable steel, it features a 0.3270 inch diameter and a 2 inch overall length. This gage is ideal for setting micrometers, NoGo inspection, and verifying other measuring devices.
  • Coating/finish: 10 Micro
  • Includes certificate of accuracy with NIST traceability
  • Suitable for setting micrometers, NoGo inspection, verniers, and other measuring devices
  • Tolerance of 0.000200 inch