Learn More
MSC Vermont Gage Class ZZ Minus Plug & Pin Gage, 0.3390 in, 2 in, Steel
Catalog No. 5000531898
Encompass
This Class ZZ Minus Plug & Pin Gage is a precision steel tool designed for accurate measurement and inspection tasks. It features a 0.3390 inch diameter and a 2 inch overall length, making it suitable for setting micrometers, performing no-go inspections, and calibrating verniers and other measuring devices.
  • Made from durable steel.
  • Features a 0.3390 inch diameter.
  • Has a 2 inch overall length.
  • Offers a tolerance of 0.000200 "
  • Includes a 10 micro coating/finish.
  • Supplied with a Certificate of Accuracy with NIST traceability.
  • Ideal for setting micrometers, no-go inspections, and calibrating measuring devices.
Catalog No. 50-005-31898 Supplier MSC Supplier No. 89033906
This Class ZZ Minus Plug & Pin Gage is a precision steel tool designed for accurate measurement and inspection tasks. It features a 0.3390 inch diameter and a 2 inch overall length, making it suitable for setting micrometers, performing no-go inspections, and calibrating verniers and other measuring devices.
  • Made from durable steel.
  • Features a 0.3390 inch diameter.
  • Has a 2 inch overall length.
  • Offers a tolerance of 0.000200 "
  • Includes a 10 micro coating/finish.
  • Supplied with a Certificate of Accuracy with NIST traceability.
  • Ideal for setting micrometers, no-go inspections, and calibrating measuring devices.