Learn More
MSC Vermont Gage Class ZZ Minus Plug & Pin Gage, 0.3430 in Diameter, 2 in OAL, Steel, Black Oxide
Catalog No. 5000529419
Encompass
This Class ZZ Minus Plug & Pin Gage is precisely engineered from steel with a black oxide finish. It has a diameter of 0.3430 inches and an overall length of 2 inches. This gage is essential for accurate measurement tasks, including setting micrometers, conducting NoGo inspections, and calibrating various measuring devices.
  • Black oxide finish
  • Constructed from steel
  • Used for setting micrometers
  • Ideal for NoGo inspection
  • Suitable for calibrating verniers and other measuring devices
  • Includes a Certificate of Accuracy with NIST traceability
Catalog No. 50-005-29419 Supplier MSC Supplier No. 08866709
This Class ZZ Minus Plug & Pin Gage is precisely engineered from steel with a black oxide finish. It has a diameter of 0.3430 inches and an overall length of 2 inches. This gage is essential for accurate measurement tasks, including setting micrometers, conducting NoGo inspections, and calibrating various measuring devices.
  • Black oxide finish
  • Constructed from steel
  • Used for setting micrometers
  • Ideal for NoGo inspection
  • Suitable for calibrating verniers and other measuring devices
  • Includes a Certificate of Accuracy with NIST traceability