Learn More
MSC Vermont Gage Class ZZ Minus Plug & Pin Gage, 0.3440 in Dia, 2 in OAL, Steel
Catalog No. 5000515201
Encompass
Vermont Gage Steel Class ZZ Minus Plug & Pin Gages are designed for making quick and accurate measurements. They are suitable for tasks such as measuring hole diameters, gaging slot widths, and setting various measuring devices. This gage features a 0.3440 inch diameter and a 2 inch overall length, manufactured to Class ZZ tolerance for precision.
  • Class ZZ tolerance of 0.0002 "
  • Constructed from tool steel with a hardness of 60/65 Rc.
  • Overall length of 2 "
  • Features a 10 microinch finish or better.
  • Round within 0.0001 "
  • Includes a certificate of accuracy with NIST traceability.
  • Ideal for setting micrometers, NoGo inspection, verniers, and other measuring devices.
  • Manufactured in the United States.
Catalog No. 50-005-15201 Supplier MSC Supplier No. 89034409
Vermont Gage Steel Class ZZ Minus Plug & Pin Gages are designed for making quick and accurate measurements. They are suitable for tasks such as measuring hole diameters, gaging slot widths, and setting various measuring devices. This gage features a 0.3440 inch diameter and a 2 inch overall length, manufactured to Class ZZ tolerance for precision.
  • Class ZZ tolerance of 0.0002 "
  • Constructed from tool steel with a hardness of 60/65 Rc.
  • Overall length of 2 "
  • Features a 10 microinch finish or better.
  • Round within 0.0001 "
  • Includes a certificate of accuracy with NIST traceability.
  • Ideal for setting micrometers, NoGo inspection, verniers, and other measuring devices.
  • Manufactured in the United States.