Learn More
MSC Vermont Gage Class ZZ Minus Plug & Pin Gage, 0.3580 in, 2 in Length
Catalog No. 5000531897
Encompass
This Class ZZ Minus Plug & Pin Gage features a 0.3580 inch diameter and a 2 inch overall length. Constructed from steel, it is designed for precise measurement applications in various settings.
  • Set micrometers, verniers, and other measuring devices
  • Suitable for NoGo inspection
  • Includes a certificate of accuracy with NIST traceability
  • Made from steel
  • Features a 10 micro coating/finish
  • Tolerance of 0.000200 inch
Catalog No. 50-005-31897 Supplier MSC Supplier No. 89035802
This Class ZZ Minus Plug & Pin Gage features a 0.3580 inch diameter and a 2 inch overall length. Constructed from steel, it is designed for precise measurement applications in various settings.
  • Set micrometers, verniers, and other measuring devices
  • Suitable for NoGo inspection
  • Includes a certificate of accuracy with NIST traceability
  • Made from steel
  • Features a 10 micro coating/finish
  • Tolerance of 0.000200 inch