Learn More
MSC Vermont Gage Class ZZ Minus Plug & Pin Gage, 0.3700 in, 2 in, Steel, 10 Micro
Catalog No. 5000531861
Encompass
This Class ZZ Minus Plug & Pin Gage is precisely engineered for accurate measurement applications. With a 0.3700 in diameter and an overall length of 2 in, it is manufactured from steel and features a 10 Micro finish. It is designed for use in setting micrometers, conducting NoGo inspections, and calibrating verniers and other measuring devices.
  • Provides a tolerance of 0.000200.
  • Made from durable steel.
  • Features a 10 Micro finish for precise measurements.
  • Suitable for setting micrometers, NoGo inspection, and calibrating verniers and other measuring devices.
  • Includes a Certificate of Accuracy with NIST Traceability.
Catalog No. 50-005-31861 Supplier MSC Supplier No. 89037006
This Class ZZ Minus Plug & Pin Gage is precisely engineered for accurate measurement applications. With a 0.3700 in diameter and an overall length of 2 in, it is manufactured from steel and features a 10 Micro finish. It is designed for use in setting micrometers, conducting NoGo inspections, and calibrating verniers and other measuring devices.
  • Provides a tolerance of 0.000200.
  • Made from durable steel.
  • Features a 10 Micro finish for precise measurements.
  • Suitable for setting micrometers, NoGo inspection, and calibrating verniers and other measuring devices.
  • Includes a Certificate of Accuracy with NIST Traceability.