Learn More
MSC Vermont Gage Class ZZ Minus Plug & Pin Gage, 0.4375 in Dia, 2 in OAL, Steel
Catalog No. 5000532330
Encompass
This Class ZZ Minus Plug & Pin Gage is designed for precise measurement and inspection tasks. Constructed from durable steel with a black oxide coating, it ensures accuracy and longevity. With a diameter of 0.4375 inch and an overall length of 2 inch, it is suitable for various calibration and inspection applications.
  • Manufactured from steel
  • Features a black oxide coating
  • Includes a certificate of accuracy with NIST traceability
  • Designed for setting micrometers
  • Suitable for NoGo inspection
  • Compatible with verniers and other measuring devices
  • Provides a measurement diameter of 0.4375 inch
  • Offers an overall length of 2 inch
  • Maintains a tolerance of 0.000200 inch
Catalog No. 50-005-32330 Supplier MSC Supplier No. 32919995
This Class ZZ Minus Plug & Pin Gage is designed for precise measurement and inspection tasks. Constructed from durable steel with a black oxide coating, it ensures accuracy and longevity. With a diameter of 0.4375 inch and an overall length of 2 inch, it is suitable for various calibration and inspection applications.
  • Manufactured from steel
  • Features a black oxide coating
  • Includes a certificate of accuracy with NIST traceability
  • Designed for setting micrometers
  • Suitable for NoGo inspection
  • Compatible with verniers and other measuring devices
  • Provides a measurement diameter of 0.4375 inch
  • Offers an overall length of 2 inch
  • Maintains a tolerance of 0.000200 inch