Learn More
MSC Vermont Gage Class ZZ Minus Plug & Pin Gage, 0.4380 in Dia, 2 in OAL
Catalog No. 5000531083
Encompass
This Vermont Gage Class ZZ Minus Plug & Pin Gage is designed for precise measurement applications such as setting micrometers, performing NoGo inspections, and calibrating verniers and other measuring devices. Constructed from steel with a 10 micro coating/finish, this gage pin ensures accuracy and durability. Each gage includes a Certificate of Accuracy with NIST Traceability.
  • Precise measurement diameter of 0.4380 in
  • Overall length of 2 in
  • Tolerance of 0.000200 in
  • Ideal for setting micrometers, NoGo inspection, and calibrating measuring devices
  • Made from durable steel
  • Features a 10 micro coating/finish
  • Includes a certificate of accuracy with NIST traceability
Catalog No. 50-005-31083 Supplier MSC Supplier No. 89043806
This Vermont Gage Class ZZ Minus Plug & Pin Gage is designed for precise measurement applications such as setting micrometers, performing NoGo inspections, and calibrating verniers and other measuring devices. Constructed from steel with a 10 micro coating/finish, this gage pin ensures accuracy and durability. Each gage includes a Certificate of Accuracy with NIST Traceability.
  • Precise measurement diameter of 0.4380 in
  • Overall length of 2 in
  • Tolerance of 0.000200 in
  • Ideal for setting micrometers, NoGo inspection, and calibrating measuring devices
  • Made from durable steel
  • Features a 10 micro coating/finish
  • Includes a certificate of accuracy with NIST traceability