Learn More
MSC Vermont Gage Class ZZ Minus Plug & Pin Gage, 0.4640 in, 2 in, Steel
Catalog No. 5000536867
Encompass
This Class ZZ Minus Plug & Pin Gage is a precision measuring tool. With a diameter of 0.4640 inches and an overall length of 2 inches, it is ideal for setting micrometers, performing NoGo inspection, and calibrating verniers and other measuring devices.
  • Includes certificate of accuracy with NIST traceability.
  • Suitable for setting micrometers and other measuring devices.
  • Designed for noGo inspection.
  • Constructed from steel for durability.
  • Features a 10 micro coating/finish.
Catalog No. 50-005-36867 Supplier MSC Supplier No. 89046403
This Class ZZ Minus Plug & Pin Gage is a precision measuring tool. With a diameter of 0.4640 inches and an overall length of 2 inches, it is ideal for setting micrometers, performing NoGo inspection, and calibrating verniers and other measuring devices.
  • Includes certificate of accuracy with NIST traceability.
  • Suitable for setting micrometers and other measuring devices.
  • Designed for noGo inspection.
  • Constructed from steel for durability.
  • Features a 10 micro coating/finish.