Learn More
MSC Vermont Gage Class ZZ Minus Plug & Pin Gage, 0.7070 in, Steel
Catalog No. 5000515694
Encompass
This Vermont Gage Class ZZ Minus Plug & Pin Gage is designed for precise measurement applications. Made from durable steel, it features a 0.7070 inch diameter and an overall length of 2 inches, making it suitable for setting micrometers, NoGo inspection, and other measuring devices.
  • Precise measurement with a 0.7070 in diameter.
  • Overall length of 2 in.
  • Durable steel construction.
  • Tolerance of 0.000200 in.
  • Features a 10 micro coating/finish.
  • Ideal for setting micrometers, NoGo inspection, and other measuring devices.
  • Includes a certificate of accuracy with NIST traceability.
Catalog No. 50-005-15694 Supplier MSC Supplier No. 89070700
This Vermont Gage Class ZZ Minus Plug & Pin Gage is designed for precise measurement applications. Made from durable steel, it features a 0.7070 inch diameter and an overall length of 2 inches, making it suitable for setting micrometers, NoGo inspection, and other measuring devices.
  • Precise measurement with a 0.7070 in diameter.
  • Overall length of 2 in.
  • Durable steel construction.
  • Tolerance of 0.000200 in.
  • Features a 10 micro coating/finish.
  • Ideal for setting micrometers, NoGo inspection, and other measuring devices.
  • Includes a certificate of accuracy with NIST traceability.