Learn More
MSC Vermont Gage Class ZZ Minus Plug & Pin Gage, 0.7230 in Dia, 2 in OAL
Catalog No. 5000514379
Encompass
This Class ZZ Minus Plug & Pin Gage, with a diameter of 0.7230 in and an overall length of 2 in, is engineered for precise measurement applications such as setting micrometers, NoGo inspection, and calibrating verniers and other measuring devices. It is constructed from steel and includes a Certificate of Accuracy with NIST Traceability.
  • Precisely measures at 0.7230 in diameter.
  • Features a 2 in overall length.
  • Offers a tight tolerance of 0.000200 in.
  • Constructed from durable steel.
  • Includes a 10 micro finish for enhanced performance.
  • Ideal for setting micrometers, NoGo inspection, and calibrating measuring devices.
  • Comes with a Certificate of Accuracy with NIST traceability.
Catalog No. 50-005-14379 Supplier MSC Supplier No. 89072300
This Class ZZ Minus Plug & Pin Gage, with a diameter of 0.7230 in and an overall length of 2 in, is engineered for precise measurement applications such as setting micrometers, NoGo inspection, and calibrating verniers and other measuring devices. It is constructed from steel and includes a Certificate of Accuracy with NIST Traceability.
  • Precisely measures at 0.7230 in diameter.
  • Features a 2 in overall length.
  • Offers a tight tolerance of 0.000200 in.
  • Constructed from durable steel.
  • Includes a 10 micro finish for enhanced performance.
  • Ideal for setting micrometers, NoGo inspection, and calibrating measuring devices.
  • Comes with a Certificate of Accuracy with NIST traceability.