Learn More
MSC Vermont Gage Class ZZ Minus Plug & Pin Gage, 0.7340 in Diameter, 2 in Overall Length, Steel
Catalog No. 5000531917
Encompass
This Class ZZ Minus Plug & Pin Gage is a precision tool for calibration and inspection. Made of steel, it has a 0.7340 in diameter and 2 in overall length, with a 0.000200 in tolerance. A 10 Micro coating/finish provides a durable surface. It includes a Certificate of Accuracy with NIST traceability, making it ideal for setting micrometers, NoGo inspection, and calibrating other measuring devices.
  • Precision measuring tool for accurate calibration and inspection.
  • Constructed from durable steel.
  • Features a 0.7340 in diameter and a 2 in overall length.
  • Maintains a tight tolerance of 0.000200 in.
  • Includes a 10 Micro coating/finish for enhanced durability.
  • Comes with a Certificate of Accuracy with NIST traceability.
  • Suitable for setting micrometers.
  • Ideal for NoGo inspection.
  • Used for calibrating verniers and other measuring devices.
Catalog No. 50-005-31917 Supplier MSC Supplier No. 89073407
This Class ZZ Minus Plug & Pin Gage is a precision tool for calibration and inspection. Made of steel, it has a 0.7340 in diameter and 2 in overall length, with a 0.000200 in tolerance. A 10 Micro coating/finish provides a durable surface. It includes a Certificate of Accuracy with NIST traceability, making it ideal for setting micrometers, NoGo inspection, and calibrating other measuring devices.
  • Precision measuring tool for accurate calibration and inspection.
  • Constructed from durable steel.
  • Features a 0.7340 in diameter and a 2 in overall length.
  • Maintains a tight tolerance of 0.000200 in.
  • Includes a 10 Micro coating/finish for enhanced durability.
  • Comes with a Certificate of Accuracy with NIST traceability.
  • Suitable for setting micrometers.
  • Ideal for NoGo inspection.
  • Used for calibrating verniers and other measuring devices.