Learn More
MSC Vermont Gage Class ZZ Minus Plug & Pin Gage, 0.75 mm, 2 in
Catalog No. 5000527139
Encompass
This Class ZZ Minus Plug & Pin Gage, manufactured from steel, features a 0.75 mm diameter and a 2 in overall length. It is designed for precise applications such as setting micrometers, NoGo inspection, verniers, and other measuring devices.
  • Includes certificate of accuracy with NIST traceability.
Catalog No. 50-005-27139 Supplier MSC Supplier No. 40428203
This Class ZZ Minus Plug & Pin Gage, manufactured from steel, features a 0.75 mm diameter and a 2 in overall length. It is designed for precise applications such as setting micrometers, NoGo inspection, verniers, and other measuring devices.
  • Includes certificate of accuracy with NIST traceability.