Learn More
MSC Vermont Gage Class ZZ Minus Plug & Pin Gage, 0.77 mm Dia, 2 in OAL
Catalog No. 5000511558
Encompass
This Class ZZ Minus Plug & Pin Gage is designed for precise measurement and inspection tasks. Made from durable steel, it features a 10 micro finish and a tight tolerance of 0.00508 mm, ensuring accuracy for critical applications.
  • For setting micrometers, verniers, and other measuring devices.
  • Suitable for no-go inspection.
  • Includes a certificate of accuracy with NIST traceability.
  • Constructed from steel for durability.
  • Features a 10 micro finish.
  • Tight tolerance of 0.00508 mm.
Catalog No. 50-005-11558 Supplier MSC Supplier No. 40428229
This Class ZZ Minus Plug & Pin Gage is designed for precise measurement and inspection tasks. Made from durable steel, it features a 10 micro finish and a tight tolerance of 0.00508 mm, ensuring accuracy for critical applications.
  • For setting micrometers, verniers, and other measuring devices.
  • Suitable for no-go inspection.
  • Includes a certificate of accuracy with NIST traceability.
  • Constructed from steel for durability.
  • Features a 10 micro finish.
  • Tight tolerance of 0.00508 mm.