Learn More
MSC Vermont Gage Class ZZ Minus Plug & Pin Gage, 0.81 mm Diameter, 2 in OAL, Steel
Catalog No. 5000529127
Encompass
This Class ZZ Minus Plug & Pin Gage is precisely engineered for accurate measurement. Featuring a 0.81 mm diameter and a 2 inch overall length, it is constructed from durable steel with a 10 micro finish. This gage is ideal for setting micrometers, NoGo inspection, and calibrating verniers and other measuring devices. Each gage includes a certificate of accuracy with NIST traceability.
  • Measures 0.81 mm in diameter
  • Features a 2 inch overall length
  • Constructed from steel
  • Has a 10 micro coating/finish
  • Provides a tolerance of 0.00508 mm
  • Suitable for setting micrometers
  • Used for NoGo inspection
  • Compatible with verniers and other measuring devices
  • Includes a certificate of accuracy with NIST traceability
Catalog No. 50-005-29127 Supplier MSC Supplier No. 40428260
This Class ZZ Minus Plug & Pin Gage is precisely engineered for accurate measurement. Featuring a 0.81 mm diameter and a 2 inch overall length, it is constructed from durable steel with a 10 micro finish. This gage is ideal for setting micrometers, NoGo inspection, and calibrating verniers and other measuring devices. Each gage includes a certificate of accuracy with NIST traceability.
  • Measures 0.81 mm in diameter
  • Features a 2 inch overall length
  • Constructed from steel
  • Has a 10 micro coating/finish
  • Provides a tolerance of 0.00508 mm
  • Suitable for setting micrometers
  • Used for NoGo inspection
  • Compatible with verniers and other measuring devices
  • Includes a certificate of accuracy with NIST traceability