Learn More
MSC Vermont Gage Class ZZ Minus Plug & Pin Gage, 0.8450 in, 2 in, Steel
Catalog No. 5000530311
Encompass
This Class ZZ Minus Plug & Pin Gage is designed for precise measurement and inspection tasks. Made of steel with a 10 Micro finish, it features a 0.8450 inch diameter and a 2 inch overall length, ensuring accuracy with a tolerance of 0.000200 decimal inches. It is supplied with a Certificate of Accuracy with NIST Traceability for assured quality.
  • Designed for setting micrometers, NoGo inspection, and use with verniers and other measuring devices.
  • Includes a Certificate of Accuracy with NIST Traceability.
  • Made of durable steel with a 10 Micro finish.
  • Precise 0.8450 inch diameter.
  • 2 inch overall length.
  • Tolerance of 0.000200 decimal inches.
Catalog No. 50-005-30311 Supplier MSC Supplier No. 78380417
Add to Cart
Add to Cart
This Class ZZ Minus Plug & Pin Gage is designed for precise measurement and inspection tasks. Made of steel with a 10 Micro finish, it features a 0.8450 inch diameter and a 2 inch overall length, ensuring accuracy with a tolerance of 0.000200 decimal inches. It is supplied with a Certificate of Accuracy with NIST Traceability for assured quality.
  • Designed for setting micrometers, NoGo inspection, and use with verniers and other measuring devices.
  • Includes a Certificate of Accuracy with NIST Traceability.
  • Made of durable steel with a 10 Micro finish.
  • Precise 0.8450 inch diameter.
  • 2 inch overall length.
  • Tolerance of 0.000200 decimal inches.