Learn More
MSC Vermont Gage Class ZZ Minus Plug & Pin Gage, 0.8470 in Dia, 2 in OAL, Steel
Catalog No. 5000516187
Encompass
The Class ZZ Minus Plug & Pin Gage is a precision steel instrument designed for accurate measurement applications. With a diameter of 0.8470 inches and an overall length of 2 inches, it is ideal for setting micrometers, conducting NoGo inspection, and calibrating verniers and other measuring devices.
  • Designed for setting micrometers, NoGo inspection, and calibrating verniers and other measuring devices.
  • Includes a certificate of accuracy with NIST traceability.
  • Constructed from steel with a 10 micro coating/finish.
Catalog No. 50-005-16187 Supplier MSC Supplier No. 89084701
The Class ZZ Minus Plug & Pin Gage is a precision steel instrument designed for accurate measurement applications. With a diameter of 0.8470 inches and an overall length of 2 inches, it is ideal for setting micrometers, conducting NoGo inspection, and calibrating verniers and other measuring devices.
  • Designed for setting micrometers, NoGo inspection, and calibrating verniers and other measuring devices.
  • Includes a certificate of accuracy with NIST traceability.
  • Constructed from steel with a 10 micro coating/finish.