Learn More
MSC Vermont Gage Class ZZ Minus Plug & Pin Gage, 0.8660 in Dia, 2 in OAL, Steel
Catalog No. 5000511218
Encompass
This Class ZZ Minus Plug & Pin Gage is designed for precise measurement and inspection applications, offering accuracy for setting various measuring devices.
  • Diameter of 0.8660 in
  • Overall length of 2 in
  • Tolerance of 0.000200 in
  • Constructed from steel
  • Features a 10 micro coating/finish
  • Suitable for setting micrometers, NoGo inspection, verniers, and other measuring devices
  • Includes a Certificate of Accuracy with NIST Traceability
  • Manufactured in the United States
Catalog No. 50-005-11218 Supplier MSC Supplier No. 89086607
This Class ZZ Minus Plug & Pin Gage is designed for precise measurement and inspection applications, offering accuracy for setting various measuring devices.
  • Diameter of 0.8660 in
  • Overall length of 2 in
  • Tolerance of 0.000200 in
  • Constructed from steel
  • Features a 10 micro coating/finish
  • Suitable for setting micrometers, NoGo inspection, verniers, and other measuring devices
  • Includes a Certificate of Accuracy with NIST Traceability
  • Manufactured in the United States