Learn More
MSC Vermont Gage Class ZZ Minus Plug & Pin Gage, 0.8900 in Dia, 2 in OAL
Catalog No. 5000516188
Encompass
This Class ZZ Minus Plug & Pin Gage is designed for precise measurement applications. It features a diameter of 0.8900 inch and an overall length of 2 inches, making it suitable for setting micrometers, NoGo inspection, verniers, and other measuring devices. Constructed from durable steel, it ensures reliable performance.
  • Provides highly accurate measurements with a tolerance of 0.000200 "
  • Constructed from durable steel for long-lasting use.
  • Ideal for setting micrometers, verniers, and other measuring equipment.
  • Suitable for NoGo inspection.
  • Includes a certificate of accuracy with NIST traceability.
  • Features a 10 micro coating/finish.
Catalog No. 50-005-16188 Supplier MSC Supplier No. 89089007
This Class ZZ Minus Plug & Pin Gage is designed for precise measurement applications. It features a diameter of 0.8900 inch and an overall length of 2 inches, making it suitable for setting micrometers, NoGo inspection, verniers, and other measuring devices. Constructed from durable steel, it ensures reliable performance.
  • Provides highly accurate measurements with a tolerance of 0.000200 "
  • Constructed from durable steel for long-lasting use.
  • Ideal for setting micrometers, verniers, and other measuring equipment.
  • Suitable for NoGo inspection.
  • Includes a certificate of accuracy with NIST traceability.
  • Features a 10 micro coating/finish.