Learn More
MSC Vermont Gage Class ZZ Minus Plug & Pin Gage, 0.9210 in, Steel
Catalog No. 5000536480
Encompass
This Class ZZ Minus Plug & Pin Gage is designed for precision measurement applications, such as setting micrometers and for NoGo inspection. It features a 0.9210 inch diameter and a 2 inch overall length, providing accurate and reliable performance for various measuring devices.
  • Precision measurement for setting micrometers, verniers, and other devices
  • Suitable for NoGo inspection
  • Constructed from steel for durability
  • Includes a certificate of accuracy with NIST traceability
  • Features a 10 micro finish
  • Diameter of 0.9210 inch
  • Overall length of 2 inch
Catalog No. 50-005-36480 Supplier MSC Supplier No. 89092100
This Class ZZ Minus Plug & Pin Gage is designed for precision measurement applications, such as setting micrometers and for NoGo inspection. It features a 0.9210 inch diameter and a 2 inch overall length, providing accurate and reliable performance for various measuring devices.
  • Precision measurement for setting micrometers, verniers, and other devices
  • Suitable for NoGo inspection
  • Constructed from steel for durability
  • Includes a certificate of accuracy with NIST traceability
  • Features a 10 micro finish
  • Diameter of 0.9210 inch
  • Overall length of 2 inch