Learn More
MSC Vermont Gage Class ZZ Minus Plug & Pin Gage, 0.9880 in Diameter, 2 in OAL, Steel
Catalog No. 5000530717
Encompass
The Vermont Gage Class ZZ Minus Plug & Pin Gage is a precision measuring tool designed for various inspection and calibration applications. Made from steel, it features a 0.9880 inch diameter and a 2 inch overall length, with a 10 Micro finish for accuracy.
  • Designed for setting micrometers
  • Suitable for NoGo inspection
  • Can be used with verniers and other measuring devices
  • Includes a certificate of accuracy with NIST traceability
  • Constructed from steel
  • Features a 10 Micro finish
  • Provides a tolerance of 0.000200 inch
Catalog No. 50-005-30717 Supplier MSC Supplier No. 89098800
The Vermont Gage Class ZZ Minus Plug & Pin Gage is a precision measuring tool designed for various inspection and calibration applications. Made from steel, it features a 0.9880 inch diameter and a 2 inch overall length, with a 10 Micro finish for accuracy.
  • Designed for setting micrometers
  • Suitable for NoGo inspection
  • Can be used with verniers and other measuring devices
  • Includes a certificate of accuracy with NIST traceability
  • Constructed from steel
  • Features a 10 Micro finish
  • Provides a tolerance of 0.000200 inch