Learn More
MSC Vermont Gage Class ZZ Minus Plug & Pin Gage, 0.9980 in Dia, 2 in OAL, Steel
Catalog No. 5000536439
Encompass
This Class ZZ Minus Plug & Pin Gage is a precision measuring tool with a 0.9980 in diameter and a 2 in overall length. Made from steel and featuring a 10 Micro finish, it is designed for applications such as setting micrometers, performing NoGo inspections, and use with verniers and other measuring devices. Each gage comes with a Certificate of Accuracy with NIST Traceability.
  • 0.9980 in diameter of measurement
  • 2 in overall length
  • 0.000200 in tolerance
  • Steel material
  • 10 Micro finish
  • Includes certificate of accuracy with NIST traceability
  • Suitable for setting micrometers, NoGo inspection, verniers and other measuring devices
  • Class ZZ Minus configuration
Catalog No. 50-005-36439 Supplier MSC Supplier No. 89099808
This Class ZZ Minus Plug & Pin Gage is a precision measuring tool with a 0.9980 in diameter and a 2 in overall length. Made from steel and featuring a 10 Micro finish, it is designed for applications such as setting micrometers, performing NoGo inspections, and use with verniers and other measuring devices. Each gage comes with a Certificate of Accuracy with NIST Traceability.
  • 0.9980 in diameter of measurement
  • 2 in overall length
  • 0.000200 in tolerance
  • Steel material
  • 10 Micro finish
  • Includes certificate of accuracy with NIST traceability
  • Suitable for setting micrometers, NoGo inspection, verniers and other measuring devices
  • Class ZZ Minus configuration