Learn More
MSC Vermont Gage Class ZZ Minus Plug & Pin Gage, 1.00 mm, 2 in, Steel
Catalog No. 5000529125
Encompass
This Class ZZ Minus Plug & Pin Gage is designed for precise measurement applications in various settings. It serves to accurately set micrometers and can be used for NoGo inspection, as well as with verniers and other measuring devices.
  • 1.00 mm diameter for accurate measurements
  • 2 in overall length for ease of use
  • 0.00508 mm tolerance for high precision
  • Constructed from durable steel material
  • Includes certificate of accuracy with NIST traceability
  • 10 micro coating/finish
Catalog No. 50-005-29125 Supplier MSC Supplier No. 40428450
This Class ZZ Minus Plug & Pin Gage is designed for precise measurement applications in various settings. It serves to accurately set micrometers and can be used for NoGo inspection, as well as with verniers and other measuring devices.
  • 1.00 mm diameter for accurate measurements
  • 2 in overall length for ease of use
  • 0.00508 mm tolerance for high precision
  • Constructed from durable steel material
  • Includes certificate of accuracy with NIST traceability
  • 10 micro coating/finish