Learn More
MSC Vermont Gage Class ZZ Minus Plug & Pin Gage, 1.12 mm Diameter, Steel, 10 Microfinish
Catalog No. 5000527020
Encompass
This Class ZZ Minus Plug & Pin Gage is engineered for precise NoGo inspection. Crafted from durable steel and featuring a 10 microfinish, it has a 1.12 mm diameter and an overall length of 2 inches. Each gage comes with a Certificate of Accuracy with NIST Traceability, ensuring reliable measurements for quality control.
  • Designed for NoGo inspection
  • Includes a Certificate of Accuracy with NIST Traceability
  • Made from steel
  • Features a 10 microfinish
  • Overall length of 2 in
  • Diameter of 1.12 mm
Catalog No. 50-005-27020 Supplier MSC Supplier No. 40428575
This Class ZZ Minus Plug & Pin Gage is engineered for precise NoGo inspection. Crafted from durable steel and featuring a 10 microfinish, it has a 1.12 mm diameter and an overall length of 2 inches. Each gage comes with a Certificate of Accuracy with NIST Traceability, ensuring reliable measurements for quality control.
  • Designed for NoGo inspection
  • Includes a Certificate of Accuracy with NIST Traceability
  • Made from steel
  • Features a 10 microfinish
  • Overall length of 2 in
  • Diameter of 1.12 mm