Learn More
MSC Vermont Gage Class ZZ Minus Plug & Pin Gage, 1.32 mm Diameter, 2 in OAL, Steel
Catalog No. 5000528884
Encompass
This Class ZZ Minus Plug & Pin Gage features a 1.32 mm diameter and a 2 in overall length, manufactured from steel. It is designed for precise measurements and is suitable for applications such as setting micrometers, NoGo inspection, and verifying verniers and other measuring devices.
  • Includes Certificate of Accuracy with NIST Traceability
  • Made from durable steel
  • Features a 1.32 mm diameter
  • Measures 2 in in overall length
  • Has a 10 Micro coating/finish
  • Ideal for setting micrometers and NoGo inspection
  • Suitable for verifying verniers and other measuring devices
Catalog No. 50-005-28884 Supplier MSC Supplier No. 40428773
This Class ZZ Minus Plug & Pin Gage features a 1.32 mm diameter and a 2 in overall length, manufactured from steel. It is designed for precise measurements and is suitable for applications such as setting micrometers, NoGo inspection, and verifying verniers and other measuring devices.
  • Includes Certificate of Accuracy with NIST Traceability
  • Made from durable steel
  • Features a 1.32 mm diameter
  • Measures 2 in in overall length
  • Has a 10 Micro coating/finish
  • Ideal for setting micrometers and NoGo inspection
  • Suitable for verifying verniers and other measuring devices