Learn More
MSC Vermont Gage Class ZZ Minus Plug & Pin Gage, 1.54 mm Diameter, Steel
Catalog No. 5000514104
Encompass
The Vermont Gage Class ZZ Minus Plug & Pin Gage is a precision measuring device designed for accurate calibration and inspection. With a diameter of 1.54 mm and an overall length of 2 in, this steel gage is suitable for various metrology applications.
  • Made from steel
  • Features a 10 micro coating/finish
  • Suitable for setting micrometers, NoGo inspection, verniers, and other measuring devices
  • Includes a certificate of accuracy with NIST traceability
  • Provides a tolerance of 0.00508 mm
  • Manufactured in the United States
Catalog No. 50-005-14104 Supplier MSC Supplier No. 40428997
The Vermont Gage Class ZZ Minus Plug & Pin Gage is a precision measuring device designed for accurate calibration and inspection. With a diameter of 1.54 mm and an overall length of 2 in, this steel gage is suitable for various metrology applications.
  • Made from steel
  • Features a 10 micro coating/finish
  • Suitable for setting micrometers, NoGo inspection, verniers, and other measuring devices
  • Includes a certificate of accuracy with NIST traceability
  • Provides a tolerance of 0.00508 mm
  • Manufactured in the United States