Learn More
MSC Vermont Gage Class ZZ Minus Plug & Pin Gage, 1.93 mm, 2 in
Catalog No. 5000529077
Encompass
This Class ZZ Minus Plug & Pin Gage is designed for precise measurement and inspection tasks. With a diameter of 1.93 mm and an overall length of 2 inches, it is suitable for setting micrometers, performing NoGo inspections, and calibrating verniers and other measuring devices. Constructed from durable steel, this gage provides reliable performance and includes a Certificate of Accuracy with NIST Traceability.
  • Designed for precise measurement and inspection tasks.
  • Features a diameter of 1.93 mm.
  • Has an overall length of 2 inches.
  • Constructed from durable steel.
  • Suitable for setting micrometers, NoGo inspection, verniers, and other measuring devices.
  • Includes a Certificate of Accuracy with NIST Traceability.
  • Features a 10 micro coating/finish.
  • Has a tolerance of 0.00508 mm.
Catalog No. 50-005-29077 Supplier MSC Supplier No. 40429383
This Class ZZ Minus Plug & Pin Gage is designed for precise measurement and inspection tasks. With a diameter of 1.93 mm and an overall length of 2 inches, it is suitable for setting micrometers, performing NoGo inspections, and calibrating verniers and other measuring devices. Constructed from durable steel, this gage provides reliable performance and includes a Certificate of Accuracy with NIST Traceability.
  • Designed for precise measurement and inspection tasks.
  • Features a diameter of 1.93 mm.
  • Has an overall length of 2 inches.
  • Constructed from durable steel.
  • Suitable for setting micrometers, NoGo inspection, verniers, and other measuring devices.
  • Includes a Certificate of Accuracy with NIST Traceability.
  • Features a 10 micro coating/finish.
  • Has a tolerance of 0.00508 mm.