Learn More
MSC Vermont Gage Class ZZ Minus Plug & Pin Gage, 10.02 mm, 2 in OAL, Steel
Catalog No. 5000527901
Encompass
This Class ZZ Minus Plug & Pin Gage is a precision measuring tool with a 10.02 mm diameter and a 2 in overall length. It is designed for accurate no-go inspection and for setting various measuring devices.
  • Made from durable steel for long-lasting performance.
  • Features a precise tolerance of 0.00508 mm.
  • Includes a certificate of accuracy with NIST traceability.
  • Suitable for setting micrometers, verniers, and other measuring devices.
  • Designed for no-go inspection of hole sizes.
  • Equipped with a 10 microinch surface finish.
  • Meets ISO 17025 standards.
Catalog No. 50-005-27901 Supplier MSC Supplier No. 40437477
This Class ZZ Minus Plug & Pin Gage is a precision measuring tool with a 10.02 mm diameter and a 2 in overall length. It is designed for accurate no-go inspection and for setting various measuring devices.
  • Made from durable steel for long-lasting performance.
  • Features a precise tolerance of 0.00508 mm.
  • Includes a certificate of accuracy with NIST traceability.
  • Suitable for setting micrometers, verniers, and other measuring devices.
  • Designed for no-go inspection of hole sizes.
  • Equipped with a 10 microinch surface finish.
  • Meets ISO 17025 standards.