Learn More
MSC Vermont Gage Class ZZ Minus Plug & Pin Gage, 13.51 mm Diameter, 2 in Overall Length, Steel
Catalog No. 5000512038
Encompass
Vermont Gage Class ZZ Minus Plug & Pin Gages are designed for precise measurement applications such as setting micrometers, NoGo inspection, and verifying other measuring devices. This particular gage features a 13.51 mm diameter and a 2 in overall length, crafted from durable steel with a 10 micro finish.
  • Includes certificate of accuracy with NIST traceability.
  • Country of origin: United States.
Catalog No. 50-005-12038 Supplier MSC Supplier No. 40440968
Vermont Gage Class ZZ Minus Plug & Pin Gages are designed for precise measurement applications such as setting micrometers, NoGo inspection, and verifying other measuring devices. This particular gage features a 13.51 mm diameter and a 2 in overall length, crafted from durable steel with a 10 micro finish.
  • Includes certificate of accuracy with NIST traceability.
  • Country of origin: United States.