Learn More
MSC Vermont Gage Class ZZ Minus Plug & Pin Gage, 14.96 mm, Steel, 10 Micro Finish
Catalog No. 5000534056
Encompass
This Class ZZ Minus Plug & Pin Gage is designed for precise NoGo inspection, featuring a 14.96 mm diameter and a 10 micro finish. Crafted from durable steel, it ensures reliable performance and comes with a Certificate of Accuracy with NIST Traceability.
  • 14.96 mm diameter
  • Class ZZ Minus
  • 0.00508 mm tolerance
  • Steel material
  • 10 Micro finish
  • 2 inch overall length
  • Ideal for NoGo inspection
  • Includes certificate of accuracy with NIST traceability
Catalog No. 50-005-34056 Supplier MSC Supplier No. 40442410
This Class ZZ Minus Plug & Pin Gage is designed for precise NoGo inspection, featuring a 14.96 mm diameter and a 10 micro finish. Crafted from durable steel, it ensures reliable performance and comes with a Certificate of Accuracy with NIST Traceability.
  • 14.96 mm diameter
  • Class ZZ Minus
  • 0.00508 mm tolerance
  • Steel material
  • 10 Micro finish
  • 2 inch overall length
  • Ideal for NoGo inspection
  • Includes certificate of accuracy with NIST traceability