Learn More
MSC Vermont Gage Class ZZ Minus Plug & Pin Gage, 15.52 mm Diameter, 2 in Length, Steel
Catalog No. 5000514099
Encompass
Vermont Gage Class ZZ Minus Plug & Pin Gages are economical, general-purpose tools designed for quick and accurate measurements. They are used to measure hole sizes, calculate distances between holes, gage slot widths, and check locations. These gages are also suitable for Go/NoGo inspection and for setting micrometers, verniers, and other measuring devices.
  • Class ZZ tolerance (0.005 mm)
  • NoGo (Minus) pin gage type
  • Made from tool steel, 60/65 Rc
  • 2 in long
  • 10 microinch finish or better
  • Round within 0.0025 mm
  • Marked with size and direction of tolerance on gages over 0.0605 in
  • Certificate of Accuracy included
  • NIST traceable
  • Individually packaged
Catalog No. 50-005-14099 Supplier MSC Supplier No. 40442972
Vermont Gage Class ZZ Minus Plug & Pin Gages are economical, general-purpose tools designed for quick and accurate measurements. They are used to measure hole sizes, calculate distances between holes, gage slot widths, and check locations. These gages are also suitable for Go/NoGo inspection and for setting micrometers, verniers, and other measuring devices.
  • Class ZZ tolerance (0.005 mm)
  • NoGo (Minus) pin gage type
  • Made from tool steel, 60/65 Rc
  • 2 in long
  • 10 microinch finish or better
  • Round within 0.0025 mm
  • Marked with size and direction of tolerance on gages over 0.0605 in
  • Certificate of Accuracy included
  • NIST traceable
  • Individually packaged