Learn More
MSC Vermont Gage Class ZZ Minus Plug & Pin Gage, 15.62 mm Dia, Class ZZ, Steel
Catalog No. 5000514010
Encompass
This Class ZZ Minus Plug & Pin Gage is designed for precision measurement and inspection tasks, such as setting micrometers, performing 'NoGo' inspections, and calibrating verniers and other measuring devices. It is an economical gage suitable for updating old sets and ensuring compliance.
  • Class ZZ tolerance of 0.005 mm
  • NoGo (Minus) tolerance direction
  • Made from durable steel
  • NIST traceable for accuracy
  • 2 inch overall length
  • 10 microinch finish or better
  • Round within 0.0025 mm
  • Includes certificate of accuracy
  • Suitable for measuring hole sizes and slot widths
  • Useful for checking locations and Go/NoGo gaging
Catalog No. 50-005-14010 Supplier MSC Supplier No. 40443079
This Class ZZ Minus Plug & Pin Gage is designed for precision measurement and inspection tasks, such as setting micrometers, performing 'NoGo' inspections, and calibrating verniers and other measuring devices. It is an economical gage suitable for updating old sets and ensuring compliance.
  • Class ZZ tolerance of 0.005 mm
  • NoGo (Minus) tolerance direction
  • Made from durable steel
  • NIST traceable for accuracy
  • 2 inch overall length
  • 10 microinch finish or better
  • Round within 0.0025 mm
  • Includes certificate of accuracy
  • Suitable for measuring hole sizes and slot widths
  • Useful for checking locations and Go/NoGo gaging