Learn More
MSC Vermont Gage Class ZZ Minus Plug & Pin Gage, 15.91 mm, 2 in, Steel
Catalog No. 5000527187
Encompass
This Class ZZ Minus Plug & Pin Gage is designed for precise measurement and inspection. It features a 15.91 mm diameter and an overall length of 2 inches, constructed from durable steel. This gage is ideal for setting micrometers, NoGo inspection, and use with verniers and other measuring devices, ensuring accuracy in various applications.
  • Designed for precise measurement and inspection
  • Constructed from durable steel
  • Includes certificate of accuracy with NIST traceability
  • Suitable for setting micrometers
  • Suitable for NoGo inspection
  • Compatible with verniers and other measuring devices
Catalog No. 50-005-27187 Supplier MSC Supplier No. 40443368
This Class ZZ Minus Plug & Pin Gage is designed for precise measurement and inspection. It features a 15.91 mm diameter and an overall length of 2 inches, constructed from durable steel. This gage is ideal for setting micrometers, NoGo inspection, and use with verniers and other measuring devices, ensuring accuracy in various applications.
  • Designed for precise measurement and inspection
  • Constructed from durable steel
  • Includes certificate of accuracy with NIST traceability
  • Suitable for setting micrometers
  • Suitable for NoGo inspection
  • Compatible with verniers and other measuring devices