Learn More
MSC Vermont Gage Class ZZ Minus Plug & Pin Gage, 16.52 mm Diameter, 2 in Overall Length, Steel
Catalog No. 5000511837
Encompass
This Class ZZ Minus Plug & Pin Gage features a 16.52 mm diameter and a 2 in overall length, manufactured from durable steel. It is designed for precise measurement and inspection tasks, ensuring accuracy with a tolerance of 0.00508 mm.
  • 16.52 mm measurement diameter
  • 2 in overall length
  • Steel construction
  • 0.00508 mm tolerance
  • 10 micro finish
  • Suitable for setting micrometers
  • Ideal for NoGo inspection
  • Compatible with verniers and other measuring devices
  • Includes a certificate of accuracy with NIST traceability
Catalog No. 50-005-11837 Supplier MSC Supplier No. 40443970
This Class ZZ Minus Plug & Pin Gage features a 16.52 mm diameter and a 2 in overall length, manufactured from durable steel. It is designed for precise measurement and inspection tasks, ensuring accuracy with a tolerance of 0.00508 mm.
  • 16.52 mm measurement diameter
  • 2 in overall length
  • Steel construction
  • 0.00508 mm tolerance
  • 10 micro finish
  • Suitable for setting micrometers
  • Ideal for NoGo inspection
  • Compatible with verniers and other measuring devices
  • Includes a certificate of accuracy with NIST traceability