Learn More
MSC Vermont Gage Class ZZ Minus Plug & Pin Gage, 17.00 mm Dia, 2 in OAL
Catalog No. 5000528140
Encompass
This Class ZZ Minus Plug & Pin Gage is engineered for precise measurement applications. With a 17.00 mm diameter and 2 inch overall length, it is crafted from steel and features a 0.00508 mm tolerance, making it suitable for critical inspection and calibration tasks.
  • Class ZZ Minus plug and pin gage
  • 17.00 mm measurement diameter
  • 2 inch overall length
  • Tolerance of 0.00508 mm
  • Constructed from steel
  • 10 Micro coating/finish
  • Designed for setting micrometers, NoGo inspection, and use with verniers
  • Includes certificate of accuracy with NIST traceability
Catalog No. 50-005-28140 Supplier MSC Supplier No. 40444457
This Class ZZ Minus Plug & Pin Gage is engineered for precise measurement applications. With a 17.00 mm diameter and 2 inch overall length, it is crafted from steel and features a 0.00508 mm tolerance, making it suitable for critical inspection and calibration tasks.
  • Class ZZ Minus plug and pin gage
  • 17.00 mm measurement diameter
  • 2 inch overall length
  • Tolerance of 0.00508 mm
  • Constructed from steel
  • 10 Micro coating/finish
  • Designed for setting micrometers, NoGo inspection, and use with verniers
  • Includes certificate of accuracy with NIST traceability