Learn More
MSC Vermont Gage Class ZZ Minus Plug & Pin Gage, 18.80 mm Diameter, 2 in Length, Steel
Catalog No. 5000528182
Encompass
This Class ZZ Minus Plug & Pin Gage is designed for precise measurement and inspection tasks. It features a specific diameter and overall length, and is constructed from durable material. The gage is suitable for setting micrometers, NoGo inspection, and use with verniers and other measuring devices.
  • Certificate of accuracy with NIST traceability included
  • 10 micro coating finish
  • Constructed from steel
  • Suitable for setting micrometers, NoGo inspection, verniers, and other measuring devices
Catalog No. 50-005-28182 Supplier MSC Supplier No. 40446254
This Class ZZ Minus Plug & Pin Gage is designed for precise measurement and inspection tasks. It features a specific diameter and overall length, and is constructed from durable material. The gage is suitable for setting micrometers, NoGo inspection, and use with verniers and other measuring devices.
  • Certificate of accuracy with NIST traceability included
  • 10 micro coating finish
  • Constructed from steel
  • Suitable for setting micrometers, NoGo inspection, verniers, and other measuring devices