Learn More
MSC Vermont Gage Class ZZ Minus Plug & Pin Gage, 18.86 mm, Steel
Catalog No. 5000528396
Encompass
This Class ZZ Minus Plug & Pin Gage is a precision measuring tool designed for various inspection and calibration tasks. Made from durable steel, it features a diameter of 18.86 mm and an overall length of 2 inches, providing reliable performance for demanding applications.
  • Includes certificate of accuracy with NIST traceability.
  • Suitable for setting micrometers.
  • Ideal for NoGo inspection.
  • Compatible with verniers and other measuring devices.
  • Crafted from steel for durability.
  • Features a 10 micro coating/finish.
Catalog No. 50-005-28396 Supplier MSC Supplier No. 40446312
This Class ZZ Minus Plug & Pin Gage is a precision measuring tool designed for various inspection and calibration tasks. Made from durable steel, it features a diameter of 18.86 mm and an overall length of 2 inches, providing reliable performance for demanding applications.
  • Includes certificate of accuracy with NIST traceability.
  • Suitable for setting micrometers.
  • Ideal for NoGo inspection.
  • Compatible with verniers and other measuring devices.
  • Crafted from steel for durability.
  • Features a 10 micro coating/finish.