Learn More
MSC Vermont Gage Class ZZ Minus Plug & Pin Gage, 2.00 mm, 2 in, Steel
Catalog No. 5000528885
Encompass
This economical Class ZZ Minus Plug & Pin Gage is designed for quick and accurate measurements. It is ideal for setting micrometers, performing NoGo inspections, and calibrating verniers and other measuring devices. Manufactured from durable steel, it features a fine finish and includes a Certificate of Accuracy with NIST Traceability.
  • Class ZZ tolerance
  • Go or NoGo inspection
  • Made from tool steel
  • 60-65 Rc hardness
  • 2 in long
  • 10 microinch finish or better
  • Round within 0.0025 mm
  • Marked with size and tolerance direction on gages over 0.0605 in
  • Certificate of Accuracy included, NIST traceable
Catalog No. 50-005-28885 Supplier MSC Supplier No. 40429458
This economical Class ZZ Minus Plug & Pin Gage is designed for quick and accurate measurements. It is ideal for setting micrometers, performing NoGo inspections, and calibrating verniers and other measuring devices. Manufactured from durable steel, it features a fine finish and includes a Certificate of Accuracy with NIST Traceability.
  • Class ZZ tolerance
  • Go or NoGo inspection
  • Made from tool steel
  • 60-65 Rc hardness
  • 2 in long
  • 10 microinch finish or better
  • Round within 0.0025 mm
  • Marked with size and tolerance direction on gages over 0.0605 in
  • Certificate of Accuracy included, NIST traceable