Learn More
MSC Vermont Gage Class ZZ Minus Plug & Pin Gage, 20.85 mm, 0.00508 mm Tolerance
Catalog No. 5000528656
Encompass
This Class ZZ Minus Plug & Pin Gage features a 20.85 mm diameter and a precise tolerance of 0.00508 mm. Constructed from durable steel, it is designed for a variety of inspection and measurement applications, ensuring accuracy and reliability in your work.
  • Measures 20.85 mm diameter
  • Has an overall length of 2 inches
  • Maintains a tight tolerance of 0.00508 mm
  • Constructed from durable steel
  • Suitable for NoGo inspection
  • Can be used to set micrometers, verniers, and other measuring devices
  • Includes a certificate of accuracy with NIST traceability
  • Features a 10 micro coating or finish
Catalog No. 50-005-28656 Supplier MSC Supplier No. 40448300
This Class ZZ Minus Plug & Pin Gage features a 20.85 mm diameter and a precise tolerance of 0.00508 mm. Constructed from durable steel, it is designed for a variety of inspection and measurement applications, ensuring accuracy and reliability in your work.
  • Measures 20.85 mm diameter
  • Has an overall length of 2 inches
  • Maintains a tight tolerance of 0.00508 mm
  • Constructed from durable steel
  • Suitable for NoGo inspection
  • Can be used to set micrometers, verniers, and other measuring devices
  • Includes a certificate of accuracy with NIST traceability
  • Features a 10 micro coating or finish