Learn More
MSC Vermont Gage Class ZZ Minus Plug & Pin Gage, 21.23 mm Diameter, 2 in OAL, 10 Microfinish
Catalog No. 5000510203
Encompass
This plug and pin gage is designed for precise dimensional checks. It features a 21.23 mm diameter and a 2 inch overall length, with a Class ZZ Minus tolerance and a 10 microfinish for high accuracy.
  • Used for inspecting, detecting, and testing applications.
  • Offers "Go/No Go" functionality for quick and accurate dimensional checks.
  • Provides high precision with a 10 microfinish.
Catalog No. 50-005-10203 Supplier MSC Supplier No. 40448680
This plug and pin gage is designed for precise dimensional checks. It features a 21.23 mm diameter and a 2 inch overall length, with a Class ZZ Minus tolerance and a 10 microfinish for high accuracy.
  • Used for inspecting, detecting, and testing applications.
  • Offers "Go/No Go" functionality for quick and accurate dimensional checks.
  • Provides high precision with a 10 microfinish.