Learn More
MSC Vermont Gage Class ZZ Minus Plug & Pin Gage, 3.22 mm Dia, 2 in OAL
Catalog No. 5000532385
Encompass
This steel Class ZZ Minus Plug & Pin Gage is designed for precision measurement applications, including setting micrometers, NoGo inspection, and calibrating verniers and other measuring devices. It comes with a Certificate of Accuracy with NIST Traceability, ensuring reliable and precise measurements.
  • Precisely measures 3.22 mm diameter.
  • Features a 2 inch overall length.
  • Constructed from durable steel.
  • Guarantees accuracy with a 0.00508 mm tolerance.
  • Ideal for setting micrometers, NoGo inspection, and calibrating various measuring devices.
  • Includes a Certificate of Accuracy with NIST Traceability.
  • Enhanced with a 10 micro coating/finish.
Catalog No. 50-005-32385 Supplier MSC Supplier No. 40430670
This steel Class ZZ Minus Plug & Pin Gage is designed for precision measurement applications, including setting micrometers, NoGo inspection, and calibrating verniers and other measuring devices. It comes with a Certificate of Accuracy with NIST Traceability, ensuring reliable and precise measurements.
  • Precisely measures 3.22 mm diameter.
  • Features a 2 inch overall length.
  • Constructed from durable steel.
  • Guarantees accuracy with a 0.00508 mm tolerance.
  • Ideal for setting micrometers, NoGo inspection, and calibrating various measuring devices.
  • Includes a Certificate of Accuracy with NIST Traceability.
  • Enhanced with a 10 micro coating/finish.