Learn More
MSC Vermont Gage Class ZZ Minus Plug & Pin Gage, 3.99 mm, 2 in
Catalog No. 5000532773
Encompass
This Vermont Gage Class ZZ Minus Plug & Pin Gage is a precision measuring tool designed for accurate calibration and inspection tasks. Made from durable steel, it features a 3.99 mm diameter and an overall length of 2 inches, making it suitable for setting micrometers, NoGo inspection, and use with verniers and other measuring devices. Each gage is finished to 10 Micro and includes a Certificate of Accuracy with NIST Traceability, ensuring reliable performance.
  • Precisely measures 3.99 mm diameter.
  • Features an overall length of 2 inches.
  • Maintains a tolerance of 0.00508 mm.
  • Constructed from durable steel.
  • Coating/finish of 10 Micro.
  • Includes certificate of accuracy with NIST traceability.
Catalog No. 50-005-32773 Supplier MSC Supplier No. 40431447
This Vermont Gage Class ZZ Minus Plug & Pin Gage is a precision measuring tool designed for accurate calibration and inspection tasks. Made from durable steel, it features a 3.99 mm diameter and an overall length of 2 inches, making it suitable for setting micrometers, NoGo inspection, and use with verniers and other measuring devices. Each gage is finished to 10 Micro and includes a Certificate of Accuracy with NIST Traceability, ensuring reliable performance.
  • Precisely measures 3.99 mm diameter.
  • Features an overall length of 2 inches.
  • Maintains a tolerance of 0.00508 mm.
  • Constructed from durable steel.
  • Coating/finish of 10 Micro.
  • Includes certificate of accuracy with NIST traceability.