Learn More
MSC Vermont Gage Class ZZ Minus Plug & Pin Gage, 4.52 mm, 2 in
Catalog No. 5000532410
Encompass
This Vermont Gage Class ZZ Minus Plug & Pin Gage features a 4.52 mm diameter of measurement and a 2-in overall length.
  • Constructed from steel
  • Tolerance of 0.00508 mm
  • Includes a certificate of accuracy with NIST traceability
  • Has a 10 micro coating/finish
  • Suitable for setting micrometers, NoGo inspection, verniers, and other measuring devices
Catalog No. 50-005-32410 Supplier MSC Supplier No. 40431975
This Vermont Gage Class ZZ Minus Plug & Pin Gage features a 4.52 mm diameter of measurement and a 2-in overall length.
  • Constructed from steel
  • Tolerance of 0.00508 mm
  • Includes a certificate of accuracy with NIST traceability
  • Has a 10 micro coating/finish
  • Suitable for setting micrometers, NoGo inspection, verniers, and other measuring devices