Learn More
MSC Vermont Gage Class ZZ Minus Plug & Pin Gage, 4.54 mm, 2 in OAL, Steel
Catalog No. 5000527141
Encompass
This Class ZZ Minus Plug & Pin Gage features a precise 4.54 mm diameter and a 2-inch overall length. Constructed from durable steel, it boasts a 10 Micro finish and a tolerance of 0.00508 mm. It is suitable for setting micrometers, NoGo inspection, and use with verniers and other measuring devices. Each gage includes a Certificate of Accuracy with NIST Traceability.
  • Precise 4.54 mm diameter with a tight tolerance of 0.00508 mm
  • Made from steel for long-lasting use
  • Ideal for setting micrometers, no-go inspection, and working with verniers and other measuring devices
  • Includes a certificate of accuracy with NIST traceability
  • Features a 10 micro finish for enhanced performance and longevity
Catalog No. 50-005-27141 Supplier MSC Supplier No. 40431991
This Class ZZ Minus Plug & Pin Gage features a precise 4.54 mm diameter and a 2-inch overall length. Constructed from durable steel, it boasts a 10 Micro finish and a tolerance of 0.00508 mm. It is suitable for setting micrometers, NoGo inspection, and use with verniers and other measuring devices. Each gage includes a Certificate of Accuracy with NIST Traceability.
  • Precise 4.54 mm diameter with a tight tolerance of 0.00508 mm
  • Made from steel for long-lasting use
  • Ideal for setting micrometers, no-go inspection, and working with verniers and other measuring devices
  • Includes a certificate of accuracy with NIST traceability
  • Features a 10 micro finish for enhanced performance and longevity